missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF, Thermo Scientific™

Product Code. 15401366
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401366 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15401366 Supplier Thermo Scientific Alfa Aesar Supplier No. H54579.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16750-63-3
Flash Point −11°C (12°F)
Molecular Formula C7H7BrMgO
MDL Number MFCD01317003
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 2-methoxyphenylmagnesium bromide, magnesium, bromo methoxyphenyl, 2-methoxyphenyl magnesium bromide, 2-methoxyphenylmagnesium bromide solution, 2-methoxyphenylmagnesiumbromide, o-methoxyphenylmagnesium bromide, bromo 2-methoxyphenyl magnesium, 2-methoxylphenylmagnesium bromide, 2-methoxyphenyl-magnesium bromide, grignard reagent
InChI Key FCPRDUXJWIUVPZ-UHFFFAOYSA-M
SMILES COC1=CC=CC=[C-]1.[Mg+2].[Br-]
IUPAC Name magnesium;methoxybenzene;bromide
Molecular Weight (g/mol) 211.341
PubChem CID 4280988
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 1.0M in 2-MeTHF
Chemical Name or Material 2-Methoxyphenylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.