Learn More
2-Methoxypropene, TRC
Product Code. 30482392
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482392 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30482392 Supplier Toronto Research Chemicals Supplier No. TRCM26505550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methoxypropene
CAS 116-11-0
Molecular Formula C4H8O
Quantity 50 g
Synonym 2-Methoxyprop-1-ene, 2-Methoxypropene, Isopropenyl Methyl Ether, Methyl 1-Methylvinyl Ether, Methyl Isopropenyl Ether
InChI Formula InChI=1S/C4H8O/c1-4(2)5-3/h1H2,2-3H3
SMILES COC(=C)C
IUPAC Name 2-methoxyprop-1-ene
Molecular Weight (g/mol) 72.11
Formula Weight 72.0575