missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,2,3,4-tetrahydroisoquinoline Hydrochloride, TRC

Product Code. 30474305
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474305 50 mg 50mg
30468247 100 mg 100mg
30467180 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474305 Supplier Toronto Research Chemicals Supplier No. TRCM22373350MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53112-33-7
Molecular Formula C10 H13 N . H Cl
Molecular Weight (g/mol) 183.68
Synonym Isoquinoline, 1,2,3,4-tetrahydro-2-methyl-, hydrochloride (1:1) (ACI), Isoquinoline, 1,2,3,4-tetrahydro-2-methyl-, hydrochloride (7CI, 9CI), 2-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, N-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, 1,2,3,4-Tetrahydro-2-methylisoquinoline hydrochloride
IUPAC Name 2-methyl-3,4-dihydro-1H-isoquinoline;hydrochloride
SMILES Cl.CN1CCc2ccccc2C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N.ClH/c1-11-7-6-9-4-2-3-5-10(9)8-11;/h2-5H,6-8H2,1H3;1H
Formula Weight 183.0815
Chemical Name or Material 2-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.