missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-cyclopentenone, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-cyclopentenone |
| CAS | 1120-73-6 |
| Molecular Formula | C6H8O |
| Quantity | 2.5 g |
| Synonym | 2-Methyl-2-cyclopentene-1-one |
| InChI Formula | InChI=1S/C6H8O/c1-5-3-2-4-6(5)7/h3H,2,4H2,1H3 |
| SMILES | CC1=CCCC1=O |
| IUPAC Name | 2-methylcyclopent-2-en-1-one |
| Molecular Weight (g/mol) | 96.13 |
| Formula Weight | 96.0575 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction