missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-2-Methyl-d3-isoborneol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | (-)-2-Methyl Isoborneol-d3 |
| CAS | 135441-89-3 |
| Molecular Formula | C11 2H3 H17 O |
| Quantity | 0.1 g |
| Synonym | (1 R,2 R,4 R)-1,2,7,7-Tetramethylbicyclo[2.2.1 ]heptan-2-ol-d3, (1 R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1 ]heptan-2-ol-d3, (-)-2-exo-Hydroxy-2-methylbornane-d3 |
| InChI Formula | InChI=1 S/C11H20O/c1-9(2)8-5-6-10(9,3)11(4,12)7-8/h8,12 H,5-7H2,1-4H3/t8-,10-,11-/m1/s1/i4D3 |
| SMILES | [2 H]C([2 H])([2 H])[C@@]1(O)C[C@H]2 CC[C@]1(C)C2(C)C |
| IUPAC Name | (1 R,3 R,4 R)-4,7,7-trimethyl-3-(trideuteriomethyl)bicyclo[2.2.1 ]heptan-3-ol |
| Molecular Weight (g/mol) | 171.29 |
| Formula Weight | 171.1702 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction