missing translation for 'onlineSavingsMsg'
Learn More

2-Methylaminopyridine, 98%, ACROS Organics™

Product Code. 10795072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10795072 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795072 Supplier Thermo Scientific Acros Supplier No. 198440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4597-87-9
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.05g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H8N2
MDL Number MFCD00006250
Quantity 5 g
Synonym 2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, methylamino-2 pyridine, n-2-pyridylmethylamine, pyridine, 2-methylamino, n-methyl-2-pyridinamine, 2-pyridinamine, n-methyl-9ci, n-methyl pyridimine
InChI Key SVEUVITYHIHZQE-UHFFFAOYSA-N
SMILES CNC1=CC=CC=N1
IUPAC Name N-methylpyridin-2-amine
Molecular Weight (g/mol) 108.14
PubChem CID 20727
Formula Weight 108.14
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Methylaminopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.