missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutyraldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-Methylbutanal |
| CAS | 96-17-3 |
| Molecular Formula | C5 H10 O |
| Quantity | 1 g |
| Synonym | Butanal, 2-methyl-, Butyraldehyde, 2-methyl- (6CI,8CI), Butyraldehyde, α-methyl- (3CI), 2-Methylbutanal, (RS)-2-Methylbutanal, (±)-2-Methylbutanal, (±)-2-Methylbutyraldehyde, 2-Ethylpropanal, 2-Formylbutane, 2-Methylbutyraldehyde, 2-Methylbutyric aldehyde, NSC 77077, α-Methyl-n-butanal, α-Methylbutanal, α-Methylbutyraldehyde, α-Methylbutyric aldehyde |
| InChI Formula | InChI=1S/C5H10O/c1-3-5(2)4-6/h4-5H,3H2,1-2H3 |
| SMILES | CCC(C)C=O |
| IUPAC Name | 2-methylbutanal |
| Molecular Weight (g/mol) | 86.13 |
| Formula Weight | 86.0732 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction