missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15430746
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430746 250 mg 250mg
15420746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430746 Supplier Thermo Scientific Alfa Aesar Supplier No. H54084.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945863-21-8
Molecular Formula C13H19BN2O3
Molecular Weight (g/mol) 262.116
MDL Number MFCD08458483
InChI Key LUMBLRWRRONQMO-UHFFFAOYSA-N
Synonym n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 2-n-methylaminocarbonyl-5-pyridineboronic acid pincol ester, 2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester, n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n-methylaminocarbonyl pyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine-2-carboxylic acid methylamide, 2-n-methylamidocarboxy-5-pyridine boronic acid pincol ester, 2-n-methylamidocarboxy-5-pyridineboronic acid pinacol ester, 2-n-methylaminocarbonyl pyridine-5-boronic acid pincol ester, 2-n-methylaminecarbonyl-5-pyridineboronic acid pinacol ester
PubChem CID 44119560
IUPAC Name N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC

Specifications

Melting Point 113°C to 115°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 262.11
Percent Purity 96%
Chemical Name or Material 2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.