missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole, 97%

Product Code. 11438926
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11438926

Brand: Thermo Scientific Alfa Aesar A17063.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-98-1 | C4H6N2 | 82.11 g/mol

2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-98-1
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
PubChem CID 12749
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1

Specifications

Melting Point 142°C to 144°C
Boiling Point 267°C to 268°C
Flash Point 155°C (311°F)
Quantity 500 g
UN Number UN3259
Beilstein 1368
Solubility Information Soluble in water and ethanol.
Formula Weight 82.11
Percent Purity 97%
Chemical Name or Material 2-Methylimidazole
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Methylimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.