missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole, TRC

Product Code. 30467863
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467863 100 g 100g
30465987 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467863 Supplier Toronto Research Chemicals Supplier No. TRCM313450100G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9 H9 N
Molecular Weight (g/mol) 131.1745
Synonym 1H-Indole, 2-methyl-, Indole, 2-methyl- (8CI), 2-Methyl-1H-indole, 2-Methylindole, NSC 7514
IUPAC Name 2-methyl-1H-indole
SMILES Cc1cc2ccccc2[nH]1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H9N/c1-7-6-8-4-2-3-5-9(8)10-7/h2-6,10H,1H3
Formula Weight 131.0735
Chemical Name or Material 2-Methylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.