missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)naphthalene, 98%

Product Code. 11480457
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480457 1 g 1g
11490457 5 g 5g
11400467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480457 Supplier Thermo Scientific Alfa Aesar Supplier No. B22368.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7433-79-6 | C11H10S | 174.261 g/mol

2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7433-79-6
Molecular Formula C11H10S
Molecular Weight (g/mol) 174.261
MDL Number MFCD00051615
InChI Key SSKUUDUKJMIOKQ-UHFFFAOYSA-N
Synonym 2-methylthio naphthalene, 2-methylmercaptonaphthalene, 2-methylsulfanyl naphthalene, 2-methylthionaphthalene, methyl2-naphthylsulfide, methyl 2-naphthyl sulfide, naphth-2-yl methyl thioether, naphthalene,2-methylthio, methyl naphthalen-2-yl sulfane
PubChem CID 139022
IUPAC Name 2-methylsulfanylnaphthalene
SMILES CSC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 62°C to 63°C
Odor Stench
Quantity 1 g
Beilstein 1859409
Solubility Information Slightly soluble in water
Formula Weight 174.27
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)naphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.