Learn More
2-(Methylthio)nicotinoyl chloride, 98%, Thermo Scientific™
Product Code. 11479036
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479036 25 g 25g
11459036 1 g 1g
11469036 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11479036 Supplier Thermo Scientific Alfa Aesar Supplier No. A17916.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97936-43-1
Molecular Formula C7H6ClNOS
Molecular Weight (g/mol) 187.64
MDL Number MFCD00051678
InChI Key OCEMBWMMHUSVMT-UHFFFAOYSA-N
PubChem CID 2774517
IUPAC Name 2-methylsulfanylpyridine-3-carbonyl chloride
SMILES CSC1=NC=CC=C1C(Cl)=O
Specifications
Melting Point 92°C to 96°C
Quantity 25 g
UN Number UN3261
Beilstein 5506864
Sensitivity Moisture sensitive
Synonym 2-methylthio nicotinoyl chloride, 2-methylthio nicotinyl chloride, 2-methylsulfanyl pyridine-3-carbonyl chloride, 2-methylthio pyridine-3-carbonyl chloride, 2-methylsulfanyl nicotinoyl chloride, 2-methylthio-3-pyridinecarbonyl chloride, 2-methylthiopyridine-3-carbonyl chloride, 3-pyridinecarbonylchloride,2-methylthio, 3-pyridinecarbonylchloride, 2-methylthio
Formula Weight 187.65
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)nicotinoyl chloride