missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Product Code. 10059361
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10059361 1 g Amber Glass Bottle 1g
10563032 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10059361 Supplier Thermo Scientific Maybridge Supplier No. CC24309DA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-33-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD03086181
InChI Key MYGVYNRBQSAMIF-UHFFFAOYSA-N
Synonym 2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
PubChem CID 2776562
SMILES OCC1=CC=CC=C1N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name [2-(morpholin-4-yl)phenyl]methanol
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (2-Morpholinophenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.