missing translation for 'onlineSavingsMsg'
Learn More

2-(N-Hexadecanoylamino)-4-nitrophenylphosphocholine Hydroxide, TRC

Product Code. 30009190
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009190 100 mg 100mg
30005256 10 mg 10mg
30008820 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009190 Supplier Toronto Research Chemicals Supplier No. TRCH293580100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60438-73-5
Molecular Formula C27 H49 N3 O7 P . H O
Molecular Weight (g/mol) 575.68
IUPAC Name 2-[[2-(hexadecanoylamino)-4-nitrophenoxy]-hydroxyphosphoryl]oxyethyl-trimethylazanium;hydroxide
SMILES [OH-].CCCCCCCCCCCCCCCC(=O)Nc1cc(ccc1OP(=O)(O)OCC[N+](C)(C)C)[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C27H48N3O7P.H2O/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27(31)28-25-23-24(29(32)33)19-20-26(25)37-38(34,35)36-22-21-30(2,3)4;/h19-20,23H,5-18,21-22H2,1-4H3,(H-,28,31,34,35);1H2
Formula Weight 575.3336
Percent Purity >95%
Chemical Name or Material 2-(N-Hexadecanoylamino)-4-nitrophenylphosphocholine hydroxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.