missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexylthiophene, 98%

Product Code. 11490503
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490503 10 g 10g
11400513 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490503 Supplier Thermo Scientific Alfa Aesar Supplier No. A12652.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18794-77-9 | C10H16S | 168.30 g/mol

2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in most organic solvents and ethanol Soluble . Insoluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18794-77-9
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.30
MDL Number MFCD00022535
InChI Key QZVHYFUVMQIGGM-UHFFFAOYSA-N
Synonym thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d
PubChem CID 87793
IUPAC Name 2-hexylthiophene
SMILES CCCCCCC1=CC=CS1

Specifications

Density 0.935
Boiling Point 228°C to 230°C
Flash Point 100°C (212°F)
Refractive Index 1.496
Quantity 10 g
Beilstein 110220
Solubility Information Soluble in most organic solvents and ethanol Soluble . Insoluble in water.
Formula Weight 168.3
Percent Purity 98%
Chemical Name or Material 2-n-Hexylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.