missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetonitrile, 97%

Product Code. 11387396
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387396 5 g 5g
11397396 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387396 Supplier Thermo Scientific Alfa Aesar Supplier No. L09384.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7498-57-9 | C12H9N | 167.21 g/mol

2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7498-57-9
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004122
InChI Key LPCWDVLDJVZIHA-UHFFFAOYSA-N
Synonym 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide
PubChem CID 82008
IUPAC Name 2-naphthalen-2-ylacetonitrile
SMILES N#CCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 82°C to 84°C
Density 1.092
Boiling Point 303°C
Quantity 5 g
UN Number UN3439
Beilstein 1862879
Solubility Information Insoluble in water.
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 2-Naphthylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.