Learn More
2-Naphthyloxyacetamide, Dr. Ehrenstorfer
Product Code. 30311470
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311470 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30311470 Supplier Dr. Ehrenstorfer Supplier No. DREC15478000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Naphthyloxyacetamide
CAS 35368-77-5
Analyte or Component Names 2-Naphthyloxyacetamide
Shipping Condition Room Temperature
Molecular Formula C12 H11 N O2
Certifications/Compliance ISO/IEC 17025
Synonym Acetamide, 2-(2-naphthyloxy)- (6CI,7CI), 2-(2-Naphthalenyloxy)acetamide, 2-(2-Naphthyloxy)acetamide, 2-Naphthyloxyacetamide
InChI Formula InChI=1S/C12H11NO2/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2,(H2,13,14)
Solution Type Neat
SMILES NC(=O)COc1ccc2ccccc2c1
IUPAC Name 2-naphthalen-2-yloxyacetamide
Industry Type Environmental Testing
Molecular Weight (g/mol) 201.22
Quantity 25 mg
Formula Weight 201.079 g/mol