missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-1H-imidazole-1-acetic Acid Methyl Ester, TRC

Product Code. 30474885
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474885 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474885 Supplier Toronto Research Chemicals Supplier No. TRCN49623510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitro-1H-imidazole-1-acetic Acid Methyl Ester
CAS 22813-31-6
Molecular Formula C6H7N3O4
Quantity 10 mg
Synonym Methyl 2-(2-Nitro-1H-imidazol-1-yl)acetate, Methyl 2-Nitroimidazole-1-acetate, 2-Nitroimidazole-1-acetic Acid Methyl Ester, NSC 302986, RGW 613
InChI Formula InChI=1S/C6H7N3O4/c1-13-5(10)4-8-3-2-7-6(8)9(11)12/h2-3H,4H2,1H3
SMILES COC(=O)Cn1ccnc1[N+](=O)[O-]
IUPAC Name methyl 2-(2-nitroimidazol-1-yl)acetate
Molecular Weight (g/mol) 185.14
Formula Weight 185.0437
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.