missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzamide, 98%

Product Code. 11489070
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489070 10 g 10g
11499070 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489070 Supplier Thermo Scientific Alfa Aesar Supplier No. A14084.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 610-15-1 | C7H6N2O3 | 166.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-15-1
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007976
InChI Key KLGQWSOYKYFBTR-UHFFFAOYSA-N
Synonym o-nitrobenzamide, benzamide, o-nitro, benzamide, 2-nitro, 2-carbamoylnitrobenzene, nitrobenzamide, nitro-benzamide, 2-nitro-benzamide, benzamide,2-nitro, benzmide, 2-nitro, wln: zvr bnw
PubChem CID 11876
IUPAC Name 2-nitrobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 174°C to 178°C
Boiling Point 317°C
Quantity 10 g
Beilstein 1950928
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.