missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-O-Methyl Guanosine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2'-O-Methyl Guanosine |
| CAS | 2140-71-8 |
| Molecular Formula | C11H15N5O5 |
| Quantity | 100 mg |
| Synonym | O2’-Methylguanosine |
| InChI Formula | InChI=1S/C11H15N5O5/c1-20-7-6(18)4(2-17)21-10(7)16-3-13-5-8(16)14-11(12)15-9(5)19/h3-4,6-7,10,17-18H,2H2,1H3,(H3,12,14,15,19)/t4-,6-,7-,10-/m1/s1 |
| SMILES | CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1n2cnc3C(=O)NC(=Nc23)N |
| IUPAC Name | 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-1H-purin-6-one |
| Molecular Weight (g/mol) | 297.27 |
| Formula Weight | 297.1073 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction