missing translation for 'onlineSavingsMsg'
Learn More

2-Octyn-1-ol, 98%

Product Code. 11377495
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g

Product Code. 11377495

Brand: Thermo Scientific Alfa Aesar L05596.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

CAS: 20739-58-6 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20739-58-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00039542
InChI Key TTWYFVOMGMBZCF-UHFFFAOYSA-N
Synonym 2-octyn-1-ol, 2-octyne-1-ol, pubchem13066, acmc-1clfg, 2-octyn 1-ol, 2-octyn-1-ol 10g
PubChem CID 140750
IUPAC Name oct-2-yn-1-ol
SMILES CCCCCC#CCO

Specifications

Density 0.88
Boiling Point 76°C to 78°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.456
Quantity 10 g
Beilstein 1744120
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 2-Octyn-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.