missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-ivabradine Hydrochloride, TRC

Product Code. 30136977
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136977 25 mg 25mg
30137266 50 mg 50mg
30006703 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136977 Supplier Toronto Research Chemicals Supplier No. TRCO85697525MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1616710-50-9
Molecular Formula C27 H34 N2 O6 . Cl H
Molecular Weight (g/mol) 519.03
Synonym 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-4,5-dihydro-7,8-dimethoxy-1H-3-benzazepine-1,2(3H)-dione Hydrochloride (1:1)
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-1,2-dihydro-3-benzazepine-4,5-dione;hydrochloride
SMILES Cl.COc1cc2C[C@H](CN(C)CCCN3CCc4cc(OC)c(OC)cc4C(=O)C3=O)c2cc1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H34N2O6.ClH/c1-28(16-19-11-18-13-23(33-3)24(34-4)14-20(18)19)8-6-9-29-10-7-17-12-22(32-2)25(35-5)15-21(17)26(30)27(29)31;/h12-15,19H,6-11,16H2,1-5H3;1H/t19-;/m1./s1
Formula Weight 518.2184
Chemical Name or Material 2-Oxo-ivabradine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.