missing translation for 'onlineSavingsMsg'
Learn More

2-(Perfluorodecyl)ethyl Acrylate, TRC

Product Code. 30134464
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134464 5 g 5g
30012527 500 mg 500mg
30003405 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134464 Supplier Toronto Research Chemicals Supplier No. TRCP2862105G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17741-60-5
Molecular Formula C15 H7 F21 O2
Molecular Weight (g/mol) 618.18
Synonym 2-Propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluorododecyl ester, Acrylic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluorododecyl ester (8CI), 1,1,2,2-Tetrahydroperfluorododecyl acrylate, 1H,1H,2H,2H-Perfluorododecyl acrylate, 2-(Perfluorodecyl)ethyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluorododecyl acrylate, Perfluorodecylethyl acrylate
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H7F21O2/c1-2-5(37)38-4-3-6(16,17)7(18,19)8(20,21)9(22,23)10(24,25)11(26,27)12(28,29)13(30,31)14(32,33)15(34,35)36/h2H,1,3-4H2
Formula Weight 618.0111
Chemical Name or Material 1H,1H,2H,2H-Perfluorododecyl acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.