missing translation for 'onlineSavingsMsg'
Learn More

2-Perfluoropropoxy-2,3,3,3-tetrafluoropropanol, TRC

Product Code. 30421334
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421334 100 mg -
30412089 500 mg 500mg
30419826 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421334 Supplier Toronto Research Chemicals Supplier No. TRCP999433100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26537-88-2
Molecular Formula C6H3F11O2
Molecular Weight (g/mol) 316.07
IUPAC Name 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propan-1-ol
SMILES OCC(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H3F11O2/c7-2(1-18,4(10,11)12)19-6(16,17)3(8,9)5(13,14)15/h18H,1H2
Formula Weight 315.9957
Chemical Name or Material 2-Perfluoropropoxy-2,3,3,3-tetrafluoropropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.