missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyphenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11328295
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328295 5 g 5g
11318295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328295 Supplier Thermo Scientific Alfa Aesar Supplier No. L10194.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25562-98-5
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00016386
InChI Key PCMQCXFXODBYGN-UHFFFAOYSA-N
Synonym 2-phenoxyphenylacetonitrile, 2-2-phenoxyphenyl acetonitrile, 2-phenoxybenzyl cyanide, benzeneacetonitrile,2-phenoxy, acmc-1cfbf, phenoxybenzeneacetonitrile, 2-phenoxyphenyl acetonitrile, 2-cyanomethyl diphenyl ether, 2-2-phenoxyphenyl ethanenitrile
PubChem CID 141221
IUPAC Name 2-(2-phenoxyphenyl)acetonitrile
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CC#N

Specifications

Density 1.11
Boiling Point 145°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.577
Quantity 5 g
Beilstein 2727468
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material 2-Phenoxyphenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.