missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-(1-piperidinyl)propane, TRC

Product Code. 30009434
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009434 100 mg 100mg
30007232 25 mg 25mg
30004383 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009434 Supplier Toronto Research Chemicals Supplier No. TRCP336010100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92321-29-4
Molecular Formula C14 H21 N
Molecular Weight (g/mol) 203.32
IUPAC Name 1-(2-phenylpropan-2-yl)piperidine
SMILES CC(C)(N1CCCCC1)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H21N/c1-14(2,13-9-5-3-6-10-13)15-11-7-4-8-12-15/h3,5-6,9-10H,4,7-8,11-12H2,1-2H3
Formula Weight 203.1674
Percent Purity >95%
Chemical Name or Material 2-Phenyl-2-(1-piperidinyl)propane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.