missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacetophenone, TRC

Product Code. 30428560
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428560 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428560 Supplier Toronto Research Chemicals Supplier No. TRCP31925050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenylacetophenone
CAS 451-40-1
Molecular Formula C14 H12 O
Quantity 50 g
Synonym 1,2-Diphenylethanone, 1,2-Diphenylethan-1-one, 1,2-Diphenylethanone, Benzyl phenyl ketone, Deoxybenzoin, Desoxybenzoin, NSC 131456, NSC 249236, NSC 6097, Phenyl Benzyl Ketone, Phenylmethyl Phenyl Ketone, α-Phenylacetophenone
InChI Formula InChI=1S/C14H12O/c15-14(13-9-5-2-6-10-13)11-12-7-3-1-4-8-12/h1-10H,11H2
SMILES O=C(Cc1ccccc1)c2ccccc2
IUPAC Name 1,2-diphenylethanone
Molecular Weight (g/mol) 196.2445
Formula Weight 196.0888
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.