missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine Hydrochloride (Phenethylamine Hydrochloride; PEA HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463946
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463946 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463946 Supplier LoGiCal Supplier No. LGCAMP1275.4701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenethylamine hydrochloride
CAS 156-28-5
Analyte or Component Names 2-Phenethylamine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H11 N . Cl H
Certifications/Compliance ISO 17034
Synonym 2-Phenylethylamine Hydrochloride, Phenethylamine Hydrochloride, PEA HCl
InChI Formula InChI=1S/C8H11N.ClH/c9-7-6-8-4-2-1-3-5-8;/h1-5H,6-7,9H2;1H
Solution Type Single Solution
SMILES Cl.NCCc1ccccc1
IUPAC Name 2-phenylethanamine;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 157.64
Quantity 1 mL
Formula Weight 157.0658 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.