Learn More
2-Pyrazinecarboxylic Acid 4-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Pyrazinecarboxylic Acid 4-Oxide |
| CAS | 874-54-4 |
| Molecular Formula | C5H4N2O3 |
| Quantity | 5 mg |
| Synonym | Acipimox Impurity 7 |
| InChI Formula | InChI=1S/C5H4N2O3/c8-5(9)4-3-7(10)2-1-6-4/h1-3H,(H,8,9) |
| SMILES | OC(=O)c1c[n+]([O-])ccn1 |
| IUPAC Name | 4-oxidopyrazin-4-ium-2-carboxylic acid |
| Molecular Weight (g/mol) | 140.1 |
| Formula Weight | 140.0222 |