missing translation for 'onlineSavingsMsg'
Learn More

2-(Pyren-1-ylaminocarbonyl)ethyl Methanethiosulfonate, TRC

Product Code. 30409855
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409855 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409855 Supplier Toronto Research Chemicals Supplier No. TRCP98975010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 384342-64-7
Molecular Formula C20 H17 N O3 S2
Quantity 10 mg
InChI Formula InChI=1S/C20H17NO3S2/c1-26(23,24)25-12-11-18(22)21-17-10-8-15-6-5-13-3-2-4-14-7-9-16(17)20(15)19(13)14/h2-10H,11-12H2,1H3,(H,21,22)
SMILES CS(=O)(=O)SCCC(=O)Nc1ccc2ccc3cccc4ccc1c2c34
IUPAC Name 3-methylsulfonylsulfanyl-N-pyren-1-ylpropanamide
Molecular Weight (g/mol) 383.48
Formula Weight 383.065
Chemical Name or Material 2-(Pyren-1-ylaminocarbonyl)ethyl Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.