missing translation for 'onlineSavingsMsg'
Learn More

2-(R)-Pentylsuccinic Acid 4-tert-Butyl Ester, TRC

Product Code. 30409546
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409546 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409546 Supplier Toronto Research Chemicals Supplier No. TRCP284550500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material 2-(R)-Pentylsuccinic Acid 4-tert-Butyl Ester
CAS 153427-69-1
Molecular Formula C13 H24 O4
Quantity 500 mg
InChI Formula InChI=1S/C13H24O4/c1-5-6-7-8-10(12(15)16)9-11(14)17-13(2,3)4/h10H,5-9H2,1-4H3,(H,15,16)/t10-/m1/s1
SMILES CCCCC[C@H](CC(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2R)-2-[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]heptanoic acid
Molecular Weight (g/mol) 244.33
Formula Weight 244.1675
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.