missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetyl chloride, 98%

Product Code. 11357346
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357346 5 g 5g
11367346 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357346 Supplier Thermo Scientific Alfa Aesar Supplier No. L09323.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 39098-97-0 | C6H5ClOS | 160.615 g/mol

2-Thiopheneacetyl chloride was used in the synthesis of (S)-ethyl-1-(2-thiopheneacetyl)-3-piperidinecarboxylate, nipecotate-containing immunopotentiator, 5-fluorouracil-cephalosporin prodrug, series of new N,N-di(thiopheneacetyl)diamines derivatives and 6-β-(thiophen-2-yl)acetamidomorphine. Also used in the synthesis of Nitrocefin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiopheneacetyl chloride was used in the synthesis of (S)-ethyl-1-(2-thiopheneacetyl)-3-piperidinecarboxylate, nipecotate-containing immunopotentiator, 5-fluorouracil-cephalosporin prodrug, series of new N,N′-di(thiopheneacetyl)diamines derivatives and 6-β-(thiophen-2′-yl)acetamidomorphine. Also used in the synthesis of Nitrocefin.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39098-97-0
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.615
MDL Number MFCD00005456
InChI Key AJYXPNIENRLELY-UHFFFAOYSA-N
Synonym 2-thiopheneacetyl chloride, 2-thiophen-2-yl acetyl chloride, 2-thienylacetyl chloride, 2-thiophene acetyl chloride, thiophene-2-acetyl chloride, 2-2-thienyl acetyl chloride, thiopheneacetyl chloride, 2-thiopheneacetylchloride, thienylacetyl chloride, thien-2-ylacetyl chloride
PubChem CID 162362
IUPAC Name 2-thiophen-2-ylacetyl chloride
SMILES C1=CSC(=C1)CC(=O)Cl

Specifications

Boiling Point 131°C to 132°C (90 mmHg)
Flash Point 101°C (213°F)
Quantity 5 g
UN Number UN3265
Beilstein 114547
Formula Weight 160.62
Percent Purity 98%
Chemical Name or Material 2-Thiopheneacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.