missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxaldehyde, TRC

Product Code. 30449745
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449745 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449745 Supplier Toronto Research Chemicals Supplier No. TRCT368100100G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiophene-2-carbaldehyde
CAS 98-03-3
Molecular Formula C5 H4 O S
Quantity 100 g
Synonym Thiophene-2-carbaldehyde, Pyrantel Embonate Imp. C (EP)
InChI Formula InChI=1S/C5H4OS/c6-4-5-2-1-3-7-5/h1-4H
SMILES O=Cc1cccs1
IUPAC Name thiophene-2-carbaldehyde
Molecular Weight (g/mol) 112.15
Formula Weight 111.9983
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.