missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneethanol Tosylate, TRC

Product Code. 30457340
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457340 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457340 Supplier Toronto Research Chemicals Supplier No. TRCT344860500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Thiopheneethanol Tosylate
CAS 40412-06-4
Molecular Formula C13 H14 O3 S2
Quantity 500 mg
Synonym 2-Thiopheneethanol 2-(4-Methylbenzenesulfonate), 2-(2-Thienyl)ethyl Toluene-p-sulfonate, 2-(2-Thienyl)ethyl Tosylate, 2-(2-Thiophene)ethanol Tosylate, 2-(Thien-2-yl)ethyl 4-Methylbenzenesulfonate, 2-Thiophene Ethanol Tosylate, 2-Thiopheneethanol p-Toluenesulfonate
InChI Formula InChI=1S/C13H14O3S2/c1-11-4-6-13(7-5-11)18(14,15)16-9-8-12-3-2-10-17-12/h2-7,10H,8-9H2,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)OCCc2cccs2
IUPAC Name 2-thiophen-2-ylethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 282.38
Formula Weight 282.0384
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.