missing translation for 'onlineSavingsMsg'
Learn More

2-(Tributylstannyl)-2-propen-1-amine, TRC

Product Code. 30457320
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize'
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457320 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30457320 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT7739705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Tributylstannyl)-2-propen-1-amine
CAS 155258-22-3
Molecular Formula C15 H33 N Sn
Quantity 5 g
InChI Formula InChI=1S/3C4H9.C3H6N.Sn/c3*1-3-4-2;1-2-3-4;/h3*1,3-4H2,2H3;1,3-4H2;
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)CN
IUPAC Name 2-tributylstannylprop-2-en-1-amine
Molecular Weight (g/mol) 346.14
Formula Weight 347.1635
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.