missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 11487677
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487677 1 g 1g
11497677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487677 Supplier Thermo Scientific Alfa Aesar Supplier No. A15710.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63968-85-4
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD00042409
InChI Key ACNBBQGAWMHXLA-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzonitrile, o-trifluoromethoxybenzonitrile, benzonitrile, 2-trifluoromethoxy, 2-trifluoromethoxy benzenecarbonitrile, pubchem4824, acmc-1b9l5, ksc493e9p, o-trifluoromethoxy benzonitrile, acnbbqgawmhxla-uhfffaoysa
PubChem CID 2777232
IUPAC Name 2-(trifluoromethoxy)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)OC(F)(F)F

Specifications

Density 1.245
Boiling Point 80°C to 82°C (15 mmHg)
Refractive Index 1.449
Quantity 1 g
UN Number UN3276
Formula Weight 187.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.