missing translation for 'onlineSavingsMsg'
Learn More

2-Vanillin, TRC

Product Code. 17912955
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17912955 Supplier Toronto Research Chemicals Supplier No. TRCV0974001G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 148-53-8
Molecular Formula C8 H8 O3
Quantity 1 g
Synonym Benzaldehyde, 2-hydroxy-3-methoxy-, m-Anisaldehyde, 2-hydroxy- (8CI), o-Vanillin (6CI), 2-Hydroxy-3-methoxybenzaldehyde, 2-Hydroxy-m-anisaldehyde, 2-Hydroxyl-3-methoxybenzaldehyde, 2-Vanillin, 3-Methoxy-2-hydroxybenzaldehyde, 3-Methoxysalicylaldehyde, 6-Formyl-2-methoxyphenol, 6-Formylguaiacol, NC 005, NSC 2150, o-Vaniline
InChI Formula InChI=1S/C8H8O3/c1-11-7-4-2-3-6(5-9)8(7)10/h2-5,10H,1H3
SMILES COc1cccc(C=O)c1O
IUPAC Name 2-hydroxy-3-methoxybenzaldehyde
Molecular Weight (g/mol) 152.15
Formula Weight 152.0473
Chemical Name or Material 2-Vanillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.