Learn More
2-Vinylpyrazine, 98%, stab. with ca 0.1% hydroquinone
Product Code. 11487866
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487866 1 g 1g
11497866 5 g 5g
11407876 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11487866 Supplier Thermo Scientific Alfa Aesar Supplier No. A14007.03
 Restricted Item
In Stock
Add to basket
CAS: 4177-16-6 | C6H6N2 | 106.13 g/mol
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Slightly soluble in methanol, water.
Notes
Store away from incompatible compounds such as oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4177-16-6
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.13
MDL Number MFCD00051591
InChI Key KANZWHBYRHQMKZ-UHFFFAOYSA-N
Synonym 2-vinylpyrazine, vinylpyrazine, pyrazine, ethenyl, pyrazine, vinyl, 2-vinyl pyrazine, unii-y6tpk237o6, ethenylpyrazine, vinyl pyrazine, 2ethenylpyrazine, acmc-20anmb
PubChem CID 77840
IUPAC Name 2-ethenylpyrazine
SMILES C=CC1=CN=CC=N1
Specifications
Density 1.041
Boiling Point 163°C to 164°C
Flash Point 60°C (140°F)
Refractive Index 1.556
Quantity 1 g
UN Number UN1993
Beilstein 105566
Solubility Information Slightly soluble in methanol,water.
Formula Weight 106.13
Percent Purity 98%
Chemical Name or Material 2-Vinylpyrazine, stabilized with ≈0.1% hydroquinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Vinylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Vinylpyrazine