Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Product Code. 15836732 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
1 x 15 mL (17 mm dia. FA)
1 x 15 mL (17 mm dia. RB)
1 x 16 mL
1 x 30 mL
1 x 50 mL (29 mm dia. FA)
1 x 50 mL (29 mm dia. RB)
2 x 15 mL
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15836732 1 x 15 mL (17 mm dia. FA) Pack of 2
15896722 1 x 15 mL (17 mm dia. RB) Pack of 2
15846732 2 x 15 mL Pack of 2
15806732 1 x 30 mL Pack of 2
15856732 1 x 16 mL Pack of 2
15816732 1 x 50 mL (29 mm dia. RB) Pack of 2
15826732 1 x 50 mL (29 mm dia. FA) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15836732 Supplier Ohaus™ Supplier No. 30314897
Only null left
Add to basket
Add to basket
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ Fixed Angle Rotors.
  • Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
  • Suitable for use in the following Frontier™ rotors:
    • R-A4x85/12MI
    • R-A6x85/13MI
    • R-A6x85/14MI
TRUSTED_SUSTAINABILITY
Specifications
Type FA
Capacity 1 x 15 mL (17 mm dia. FA)
Max. Tube Diameter 17 mm
For Use With (Equipment) R-A4x85/12MI, R-A6x85/13MI, R-A6x85/14MI Rotors
Description Adapter, 1x15ml D17mm FA, 2/pk
No. of Places 1
No. per Pack 2