Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ R-A6x250/8MI Fixed Rotors
Product Code. 15886642 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
1 x 50 mL (29 mm dia.)
1 x 50 mL (30 mm dia)
2 x 30 mL
4 x 15 mL
5 x 10 mL
8 x 1.5 mL
Unit Size:
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15886642 8 x 1.5 mL Pack of 2
15896642 5 x 10 mL Pack of 2
15816652 2 x 30 mL Pack of 2
15806652 4 x 15 mL Pack of 2
15826652 1 x 50 mL (29 mm dia.) Pack of 2
15836652 1 x 50 mL (30 mm dia) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15886642 Supplier Ohaus™ Supplier No. 30304369
Only null left
Add to basket
Add to basket
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-A6x250/8MI Fixed Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-A6x250/8MI Fixed Rotors
TRUSTED_SUSTAINABILITY
Specifications
Capacity 8 x 1.5 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) R-A6x250/8MI Fixed Rotors
Description Adapter, 8x1.5ml D11mm, 2/pk
No. of Places 8
No. per Pack 2