missing translation for 'onlineSavingsMsg'
Learn More

200.7, 6010 - Interference Check Solution 4: Na @ 1000; Al @ 1200; Mg @ 3000; Fe @ 5000; Ca @ 6000 μg/mL in 5% HNO3, VHG™

Product Code. 17999279
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17999279 500 mL 500mL
17938889 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17999279 Supplier VHG Labs Supplier No. VHGINT4500

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

200.7, 6010 - Interference Check Solution 4: Na @ 1000; Al @ 1200; Mg @ 3000; Fe @ 5000; Ca @ 6000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium/Aluminium/Magnesium/Iron/Calcium
CAS 7440-23-5, 7429-90-5, 7439-95-4, 7439-89-6, 7440-70-2
Methods ICP
Analyte or Component Names Sodium/Aluminium/Magnesium/Iron/Calcium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Sodium 1000 μg/mL/Aluminium 1200 μg/mL/Magnesium 3000 μg/mL/Iron 5000 μg/mL/Calcium 6000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.