Learn More
Value Plastics 200 Series Hose Barb Fittings, Elbow
Product Code. 30053622 Shop All Cole-Parmer Products
Change view
Click to view available options
Packaging Quantity:
25/Pk.
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
30053622 25/Pk. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30053622 Supplier Value Plastics Supplier No. L230J2A
  • ONLINE EXCLUSIVE PRICE
    65.35€
    57.15€ / Pack of 25
    Save 8.20€
    12% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A secure connection under pressure with 200 series barbs
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of fle in., ible tubing. Constructed of USP compliant PVDF
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Packaging Quantity 25/Pk.
Outlet Port Hose Barb Union
Sterilization Method Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave
Shape Elbow
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.