Learn More
Invitrogen™ 20S Proteasome Monoclonal Antibody (863437)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA562036
Product Code. 30536232
  • 469.00€ / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The 20S proteasome is a core component of the proteasome complex, which plays a crucial role in the degradation of intracellular proteins. This multi-subunit protease complex is essential for maintaining cellular homeostasis by selectively degrading misfolded, damaged, or unneeded proteins, thus regulating various cellular processes, including the cell cycle, signaling pathways, and stress responses. The 20S proteasome is composed of 28 subunits arranged in a barrel-shaped structure with two outer alpha-rings and two inner beta-rings, which house the proteolytic active sites. Proteins targeted for degradation are typically tagged with ubiquitin molecules, recognized by the regulatory 19S cap complex, and then translocated into the 20S core for proteolysis. The proper functioning of the 20S proteasome is vital for cellular health, and its dysfunction is associated with a range of diseases, including neurodegenerative disorders like Alzheimer's and Parkinson's disease, where protein aggregation occurs, and cancer, where proteasome inhibitors are used as therapeutic agents. Understanding the structure, function, and regulation of the 20S proteasome is key for developing new therapeutic strategies aimed at modulating proteasome activity for the treatment of various diseases. Research into the 20S proteasome continues to provide insights into its broader role in cellular physiology and its potential as a target for drug development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20S Proteasome
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Human 20S Proteasome purified from human erthrocytes.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot, Immunocytochemistry
863437
Unconjugated
Mouse
Protein A/G
RUO
Human
Antibody
IgG2b