missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL3 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67733-1-IG-20UL
This item is not returnable.
View return policy
Description
The METTL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human METTL3. This antibody recognizes human, mouse antigen. The METTL3 antibody has been validated for the following applications: WB, IHC, IF-P, FC (Intra), ELISA analysis.Specifications
| METTL3 | |
| Monoclonal | |
| 1000 μg/mL | |
| IME4; M6A; MGC4336; MT-A70; Spo8 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 65-70 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 2D8H1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 56339 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction