Learn More
Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet With Manual-Closing Doors
GREENER_CHOICE
Product Code. 11965426
Change view
Click to view available options
Door Style:
Manual-close
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Unit Size
11965426 Manual-close Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11965426 Supplier Justrite™ Supplier No. 8922021
  • ONLINE EXCLUSIVE PRICE
    2451.75€
    1977.14€ / Pack of 1
    Save 474.61€
    19% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet With Manual-Closing Doors is specially designed for safely storing acids and corrosives in laboratories.
Justrite blue steel Slimline safety cabinets for corrosives have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet. Manual-close door(s) opens to a full 180 degrees and self-latches when pushed closed.
Specifications
Description Cold-rolled Steel (18 ga)
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Door Style Manual-close
Load Bearing Capacity 22 gal.
Color Blue
Material Steel (CR)