Learn More
Hamilton™ 22 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
Product Code. 12361808
Change view
Click to view available options
Needle Gauge:
22 G
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
12361808 22 G Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12361808 Supplier Hamilton™ Supplier No. 2202202
  • ONLINE EXCLUSIVE PRICE
    135.00€
    126.50€ / Pack of 6
    Save 8.50€
    6% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Specification Sheet
  • Gauge – 22 gauge
  • Point Style – 5 or AS
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – No Hub
Specifications
Needle Gauge 22 G
Product Type Hubless Needles