missing translation for 'onlineSavingsMsg'
Learn More

25-Hydroxy Vitamin D3 3,3'-Biotinylaminopropyl Ether, TRC

Product Code. 30004857
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004857 2.5 mg 2.5mg
30135730 10 mg 10mg
30138635 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004857 Supplier Toronto Research Chemicals Supplier No. TRCH9958152.5MG
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193278-59-0
Molecular Formula C40H65N3O4S
Molecular Weight (g/mol) 684.04
Synonym [3aS-(3aalpha,4beta,6aalpha)]-Hexahydro-N-[3-[[(3beta,5Z,7E)-25-hydroxy-9,10-secocholesta-5,7,10(19)-trien-3-yl]oxy]propyl]-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
IUPAC Name N-(3-(((1S,Z)-3-(2-((1S,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexyl)oxy)propyl)-5-((3aS,4R,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide
SMILES OC(C)(CCC[C@H]([C@H](CCC1/2)[C@@]1(C)CCCC2=C/C=C3C[C@@H](OCCCNC(CCCC[C@@H]4[C@@]5([C@]([H])(CS4)NC(N5)=O)[H])=O)CCC/3=C)C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C40H65N3O4S/c1-27-15-18-31(47-24-10-23-41-36(44)14-7-6-13-35-37-34(26-48-35)42-38(45)43-37)25-30(27)17-16-29-12-9-22-40(5)32(19-20-33(29)40)28(2)11-8-21-39(3,4)46/h16-17,28,31-35,37,46H,1,6-15,18-26H2,2-5H3,(H,41,44)(H2,42,43,45)/b29-16+,30-17-/t28-,31+,32+,33?,34+,35-,37+,40-/m1/s1
Formula Weight 683.47
Chemical Name or Material 25-Hydroxy Vitamin D3 3,3'-Biotinylaminopropyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.