Learn More
25-OH Vitamin D3 Antibody-antigen Complex Antibody (RM428) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-26039JF635
Product Code. 30176182
  • 683.00€ / 0.10mL
Description
25-OH Vitamin D3 Antibody-antigen Complex Monoclonal antibody specifically detects 25-OH Vitamin D3 Antibody-antigen Complex in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25-OH Vitamin D3 Antibody-antigen Complex
Monoclonal
Janelia Fluor 635
Rabbit
Protein A purified
RUO
Non-species specific
Purified
ELISA
RM428
50mM Sodium Borate
25-OH Vitamin D3 Antibody-antigen Complex
0.1 mL
Primary
Store at 4°C in the dark.