missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine Clonixinate, Mikromol™
Description
Lysine Clonixinate
Specifications
Specifications
| Chemical Name or Material | Lysine Clonixinate |
| CAS | 55837-30-4 |
| Analyte or Component Names | Lysine Clonixinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H11 Cl N2 O2 . C6 H14 N2 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | L-Lysine, 2-[(3-chloro-2-methylphenyl)amino]-3-pyridinecarboxylate (1:1), L-Lysine, mono[2-[(3-chloro-2-methylphenyl)amino]-3-pyridinecarboxylate] (9CI), 3-Pyridinecarboxylic acid, 2-[(3-chloro-2-methylphenyl)amino]-, compd. with L-lysine (1:1) (9CI), Clonix, Clonixin lysinate, Clonixin lysine salt, Deltar, Dolagial, Dorixina, L 104, L 104 (pharmaceutical), Lysine clonixinate |
| InChI Formula | InChI=1S/C13H11ClN2O2.C6H14N2O2/c1-8-10(14)5-2-6-11(8)16-12-9(13(17)18)4-3-7-15-12;7-4-2-1-3-5(8)6(9)10/h2-7H,1H3,(H,15,16)(H,17,18);5H,1-4,7-8H2,(H,9,10)/t;5-/m.0/s1 |
| SMILES | Cc1c(Cl)cccc1Nc2ncccc2C(=O)O.NCCCC[C@H](N)C(=O)O |
| IUPAC Name | 2-(3-chloro-2-methylanilino)pyridine-3-carboxylic acid;(2S)-2,6-diaminohexanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?