missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyltryptamine Oxide, TRC

Product Code. 17969754
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969754 250 mg 250mg
30005651 500 mg 500mg
17949454 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17969754 Supplier Toronto Research Chemicals Supplier No. TRCD471625250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948-19-6
Molecular Formula C12 H16 N2 O
Molecular Weight (g/mol) 204.27
Synonym N,N-Dimethyltryptamine N-oxide, N,N-dimethyl N-oxide-1H-indole-3-ethanamine
IUPAC Name 2-(1H-indol-3-yl)-N,N-dimethyl-ethanamine oxide
SMILES CN(=O)(C)CCc1c[nH]c2ccccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H16N2O/c1-14(2,15)8-7-10-9-13-12-6-4-3-5-11(10)12/h3-6,9,13H,7-8H2,1-2H3
Formula Weight 204.1263
Percent Purity >95%
Chemical Name or Material N,N-DMT N-oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.